Order Entry
Belgium
ContactUsLinkComponent
 
 
Sodium hydroxide 1 mol/l (1 N) in aqueous solution
  FFCHFF1269010
 :  
 
Sodium hydroxide 1 mol/l (1 N) in aqueous solution
  FFCHFF1269010
 :  

 

  • Pk:
    10 L
  • Seq:
    5214

 

 

 :
 :40 g/mol
 :1.04 g/cm³ (25 °C)
 :Ambient
 :215-185-5
 :3266
 :8,III
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548