Sodium hydroxide 0.1 mol/l (0.1 N) in aqueous solution
Catalog # FFCHFF60160500
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:40 g/mol
Density:1 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,III
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548