Order Entry
Belgium
ContactUsLinkComponent
Sodium hydroxide ≥98.0% (by acidimetry), pellets, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Corrosive
Sodium hydroxide ≥98.0% (by acidimetry), pellets, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Corrosive
Sodium hydroxide ≥98.0% (by acidimetry), pellets, BAKER ANALYZED®, Electronic Grade for microelectronic, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Microelectronic Use:
    = 98.0 %
  • Assay (NaOH) (by acidimetry):
    ≥98.0 %
  • Sodium Carbonate (Na₂CO₃):
    ≤0.4 %
  • Insoluble Matter:
    ≤30 ppm
  • Ammonium Hydroxide Precipitate:
    ≤0.01 %
  • Potassium (K)(by FES):
    ≤0.02 %
  • Nitrogen Compounds (as N):
    ≤3 ppm
  • Chloride (Cl):
    ≤0.005 %
  • Phosphate (PO₄):
    ≤2 ppm
  • Sulfate (SO₄):
    ≤30 ppm
  • Trace Impurities - Copper (Cu):
    ≤10 ppm
  • Heavy Metals (as Ag):
    ≤10 ppm
  • Trace Impurities - Iron (Fe):
    ≤3.000 ppm
  • Trace Impurities - Mercury (Hg)(by AAS):
    ≤0.100 ppm
  • Trace Impurities - Nickel (Ni):
    ≤5.000 ppm

Specification Test Results

Related Information

Frequently Bought Together