Order Entry
Belgium
ContactUsLinkComponent
 
Disodium hydrogen phosphate heptahydrate 98.0-100.5% (dried basis), crystals USP endotoxin tested, J.T.Baker®
 :  Avantor
 
undefined
Disodium hydrogen phosphate heptahydrate 98.0-100.5% (dried basis), crystals USP endotoxin tested, J.T.Baker®
 :  Avantor
 

 

  • GMP Manufactured Product:
    98.0 - 100.5%
  • Meets U.S.P Requirements:
    ≤16 ppm
  • CAUTION: For Manufacturing, processing or repackaging:
    ≤0.06%
  • Bulk Pharmaceutical Chemical:
    Passes Test
  • USP - Assay (Na₂HPO₄) (dried basis):
    98.0 - 100.5 %
  • USP - Arsenic (As):
    ≤ 16 ppm
  • USP - Chloride (Cl):
    ≤ 0.06 %
  • USP - Endotoxin Concentration (2.5 EU/g max.):
    Passes Test
  • USP - Identification:
    Passes Test
  • USP - Insoluble Substances:
    ≤ 0.4 %
  • USP - Loss on Drying:
    43.0 - 50.0 %
  • USP - Sulfate (SO₄):
    ≤ 0.2 %

 

 

Frequently Bought Together