Sodium dihydrogen phosphate monohydrate 98.0-103.0% (by anhydrous basis), crystals USP, FCC endotoxin tested, J.T.Baker®
: Avantor
: Sodium phosphate monobasic monohydrate, Sodium dihydrogen orthophosphate monohydrate, Monosodium phosphate monohydrate, Sodium dihydrogenorthophosphate monohydrate, Sodium orthophosphate monobasic monohydrate
:10049-21-5
:
:NaH₂PO₄·H₂O
:MFCD00149208
:137.99 g/mol
:13,08734
:100 °C
:2.04 g/cm³ (20 °C)
:Ambient
:231-449-2
:01-2119489796-13
- Environmentally Preferable:
- Meets F.C.C. Requirements:98.0 - 103.0%
- Meets U.S.P Requirements:Passes Test
- GMP Manufactured Product:≤3 ppm
- Food GMP Manufactured Product:≤0.014%
- CAUTION: For Manufacturing, processing or repackaging:Passes Test
- Bulk Food Chemical:Passes Test
- Bulk Pharmaceutical Chemical:≤0.2%
- USP - Assay (anhydrous basis):98.0 - 103.0 %
- USP - Aluminum, Calcium, and Related Elements:Passes Test
- USP - Arsenic (As):≤3 ppm
- USP - Chloride (Cl):≤0.014 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Insoluble Substances:≤0.2 %
- USP - pH (1 in 20):4.1 - 4.5
- USP - Sulfate (SO₄):≤0.15 %
- USP - Water (H₂O):10.0 - 15.0 %
- FCC - Assay (anhydrous basis):98.0 - 103.0 %
- FCC - Arsenic (As):≤3 ppm
- FCC - Fluoride (F):≤0.005 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Insoluble Substances:≤0.2 %
- FCC - Lead (Pb):≤4 mg/kg
- FCC - Loss on Drying:10.0 - 15.0 %
- Endotoxin Concentration (EU/g):≤2.5
Frequently Bought Together