0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Iodine
Iodine 0.05 mol/l (0.1 N) in aqueous solution
Iodine 0.05 mol/l (0.1 N) in aqueous solution
FFCHFF0670500
:
FF CHEMICALS
:
Iodine 0.05 mol/l (0.1 N) in aqueous solution
FFCHFF0670500
:
FF CHEMICALS
:
Pack type:
Glass bottle
Pk:
500 mL
Seq:
2725
:
:
253.81 g/mol
:
Ambient
:
231-442-4
:
3264
:
8,II
:
7553-56-2
:
I₂
:
MFCD00011355