Allyl hexanoate, Sigma-Aldrich®
8.14990.0250
: Merck
:
- Pack type:Glass bottle
- Pk:250 ml
- Seq:2590
- Environmentally Preferable:
- Solubility:0.06 g/l (20 °C)
- Molar Mass:156.23 g/mol
- Boiling Point:186 - 188 °C
- Flash Point:74 °C
- Density:0.887 g/cm3 (25 °C)
: Hexanoic acid allyl ester
:
:156.22 g/mol
:75 °C (1013 hPa)
:0.8886 g/cm³ (20 °C)
:6.1,III
:M-Clarity™ quality level = MQ200
:204-642-4
:2810
:GHS06
:GHS09
:Ambient
:123-68-2
:C₉H₁₆O₂
:MFCD00038339