
Acetic acid 2 mol/l (2 N)
Catalog # FFCHFF6549010
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pk:10 L
- Seq:5214
Specifications
About this item
:
:60.05 g/mol
Storage Temperature:Ambient
EINECS:200-580-7
UN:2790
ADR:8,III
:GHS07
CAS Number:64-19-7
Formula:H₃CCOOH
MDL Number:MFCD00036152