

Sodium acetate 0.1 M CH3COONa in acetic acid volumetric solution, Fluka™
Catalog # 35208-1L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
Specification Test Results
- Factor:0.99 - 1.01
Specification Test Results
About this item
:
:82.03 g/mol
Boiling Pt:117-118 °C
Boiling Pt:117…118 °C
Melting Pt:16.2 °C
Density:1.05 g/cm³
Flash Pt:19.9 %(V)
Storage Temperature:Ambient
EINECS:200-580-7
UN:2789
ADR:8,II
:GHS02
:GHS05
CAS Number:127-09-3
Formula:H₃CCOONa
MDL Number:MFCD00137248