ContactUsLinkComponent
Ammonium heptamolybdate tetrahydrate
  FFCHFF60960050
 :  
Ammonium heptamolybdate tetrahydrate
  FFCHFF60960050
 :  
  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370
 :
 :1235.86 g/mol
 :13,00536
 :2.498 g/cm³ (20 °C)
 :234-722-4
 :GHS07
 :Ambient
 :12054-85-2
 :12027-67-7
 :(NH₄)₆Mo₇O₂₄·4H₂O
 :MFCD00167059