Order Entry
Belgium
Orders LinkContactUsLinkComponent
Ammonia solution 10%

Ammonia solution 10%

10 L
Catalog # FFCHFF20209010
Corrosive
Irritant
Environment
Supplier:   FF CHEMICALS
Ammonia solution 10%
Catalog # FFCHFF20209010
Supplier:  FF CHEMICALS
CAS Number:  1336-21-6
Ammonia solution 10%

10 L

CAS Number:  1336-21-6
Price
Inquire For Price
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.92 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specifications

  • Pk:
    10 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.