Ammonia solution 0.1 mol/l (0.1 N) volumetric solution
Catalog # REGENH20101
Supplier: REAGECON
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
About this item
Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
:
:35.05 g/mol
Storage Temperature:Ambient
EINECS:215-647-6
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650