Piperidin-4-ol 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 4-Piperidinol, 4-Hydroxypiperidine
CAS Number:5382-16-1
:
Formula:C₅H₁₁NO
MDL Number:MFCD00005999
Boiling Pt:108…114 °C (10 mmHg)
Molecular Weight:101.15 g/mol
Melting Pt:85…90 °C
Flash Pt:107 °C
Storage Temperature:Ambient
EINECS:226-373-1
UN:3263
ADR:8 II
:GHS07
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:85 to 90 °C
- Water:≤1 % (K.F.)
- GC:≥99.0 %
Specification Test Results
Frequently Bought Together