Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, 2-Hexanone
Supelco® Organic Reference Standard, 2-Hexanone
Catalog # 47733-U
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, 2-Hexanone
Catalog # 47733-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Standards:
    Analytical standard
  • CAS No.:
    591-78-6
  • Description:
    2-Hexanone
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H226 - H336 - H361f - H372
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00009482
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    Butyl methyl ketone
  • Melting point:
    –57 °C (lit.)
  • Formula:
    CH₃(CH₂)₃COCH₃
  • Molecular weight:
    100,16
  • Density:
    0,812 g/ml at 25 °C (lit.)
  • Flash point:
    23 °C
  • Boiling point:
    127 °C (lit.)

Specifications

About this item

  • GHS signal: GHS02, GHS08, GHS07
Caution: P202 - P210 - P233 - P240 - P241 - P308 + P313