Order Entry
Belgium
ContactUsLinkComponent
m-Xylene
Flammable
Irritant
m-Xylene
Catalog # FFCHFF20321000
Supplier:  FF CHEMICALS
CAS Number:  
Flammable
Irritant
m-Xylene
Catalog # FFCHFF20321000
Supplier:  FF CHEMICALS
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :106.17 g/mol
Merck Index:12,10214
Boiling Pt:138 °C (1013 hPa)
Melting Pt:–48 °C
Density:0.864 g/cm³ (20 °C)
Flash Pt:25 °C
Storage Temperature:Ambient
EINECS:203-576-3
UN:1307
ADR:3,III
 :GHS02
 :GHS07
CAS Number:108-38-3
Formula:C₈H₁₀
MDL Number:MFCD00008536