Order Entry
Belgium
ContactUsLinkComponent
2,6-Di-tert-butyl-p-cresol
2,6-Di-tert-butyl-p-cresol
Catalog # PROOLGC7300
CAS Number:  
2,6-Di-tert-butyl-p-cresol
Catalog # PROOLGC7300
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :220.35 g/mol
Merck Index:13,01547
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-881-4
CAS Number:128-37-0
Formula:C₁₅H₂₄O
MDL Number:MFCD00011644