Orders LinkContactUsLinkComponent
Molecular model: electron repulsion
Molecular model: electron repulsion
Molecule model: Electron repulsion
Catalog # 761-0116
Marketsource
Supplier:   SPIRING ENTERPRISES
Molecular model: electron repulsion
Catalog # 761-0116
Marketsource
Molecular model: electron repulsion
Molecular model: electron repulsion
Molecule model: Electron repulsion
Price
20,50 €
Log in for availability
1 * 1 SET
Product Details & Documents
Lone pairs of electrons are represented with brown balls.
Two additional pear-shaped electron clouds are included to demonstrate the hydronium ion that forms from a hydrogen ion from hydrochloric acid.
Ordering information: Contains components for building eight models illustrating bonds in hydrochloric acid (straight), beryllium chloride (straight), water (angled), boron trihydride (flat trigonal), ammonia (pyramidal), methane (tetrahedral), phosphorus pentachloride (trigonal bipyramidal) and sulfur hexafluoride (octahedral).
Specifications
  • Description:
    Molecule model: Electron repulsion