Orders LinkContactUsLinkComponent
Orbital molecular models
Orbital molecular models
Orbital molecular models
Catalog # 761-0114
Marketsource
Supplier:   SPIRING ENTERPRISES
Orbital molecular models
Catalog # 761-0114
Marketsource
Orbital molecular models
Orbital molecular models
Orbital molecular models
Price
38,44 €
Log in for availability
1 * 1 SET
Product Details & Documents
Four models of molecular orbital configurations using benzene, ethane, ethene, and ethyne.
The models depict sigma and pi bonds and effects such as hybridisation and delocalised electrons.
Specifications
  • Description:
    Orbital molecular models