Order Entry
Belgium
ContactUsLinkComponent
 
GranuCult® plus m-TGE (modified Tryptone Glucose Meat extract) Broth
Catalog # 1.17337.0500
Supplier:  Merck
CAS Number:  
GranuCult® plus m-TGE (modified Tryptone Glucose Meat extract) Broth
Catalog # 1.17337.0500
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 g
  • Description:
    m-TGE (modified Tryptone Glucose Meat extract) broth
  • Storage temperature:
    15...25 °C

Specifications

About this item

This medium is recommended for total heterotrophic microorganism detection in water and other liquid samples.

Tryptone Glucose Extract Broth (TGE) is a non-selective nutrient medium and typically used to detect total heterotrophic microorganisms in water, dairy products, and other products like beer, bottled water, cider, soft drinks, and wine samples by membrane filtration technique. It is used where inhibitors are not present. TGE is used as a standard medium for the bacteriological plate count and can also be used for the detection of thermophilic bacteria in milk and dairy products. Tryptone and meat extract provide nitrogenous and carbonaceous compounds, long chain amino acids, vitamin B complex and other essential growth nutrients. Glucose is the energy source.

Typical composition: Enzymatic digest of casein (Tryptone), Meat extract and D(+)Glucose (Dextrose).

Pack stored upright, protected from light and properly sealed. After first opening of the bottle the content can be used up to the expiry date when stored dry and tightly closed.