Snooded Polypropylene Caps
Snooded PP caps, White
Catalog # HOPE70.401
Supplier: Medicom
Jump to:
Product Details & Documents
These snooded caps provide extra protection for workers with long hair in industrial environments.
- Fully protects and contains long hair
- Conveys professional image
- Ability to use color code for tracking
- Single use
Delivery information: Includes caps and dispenser box.
Documents
TDS PP Snooded Peaked Cap.pdf
Datasheet
Specifications
- Colour:White
- Description:Snooded PP caps
- Cat. No.:HOPE70.401
- Garment application:Laboratory|Industrial
- Weight:40 g/m²