Orders LinkContactUsLinkComponent
Refractix™ Universal Digital Refractometer
Refractix™ Universal Digital Refractometer
Digital refractometer with color touch screen
Catalog # REIC13910000
Supplier:   REICHERT
Refractix™ Universal Digital Refractometer
Catalog # REIC13910000
Supplier:  REICHERT
Refractix™ Universal Digital Refractometer
Refractix™ Universal Digital Refractometer
Digital refractometer with color touch screen
Price
2 790,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Refractometer provides laboratory performance in a compact portable platform.
  • Sapphire prism
  • 250 measurement scale capacity with 50 scales pre-loaded
  • 2,8" color touch screen and physical buttons
  • High resolution 1500 CCD linear image sensor with benchtop accuracy
  • Measurement log stores last 5000 measurements
  • Bluetooth and USB-C connectivity to RefractixSync PC software
  • Rechargeable battery with AC power option
  • Designed, engineered, and manufactured in the USA
The Refractix™ refractometer is innovative, thoughtfully designed, portable, and defies classification. Can be used in the laboratory with benchtop accuracy/precision and can be used as a portable on the go. 250 measurement scale capacity with 50 scales preloaded and found in scale groups for food/beverage, chemical, beer/wine/spirits, automotive, veterinary and favorites.
View real time measurements and download 5000 measurement log through RefractixSync software PC based App. Laboratory performance in a value added and affordable platform.
Specifications
  • Description:
    Digital refractometer with color touch screen
  • Measurement range:
    1,3300 - 1,5600 nD (0,0 - 85,0/100 g Brix ICUMSA)
  • Cat. No.:
    REIC13910000
  • Accuracy:
    0,0001 nD (0,1/100 g brix) [precision]
  • W×D×H:
    17,6×7,3×4,2 cm
  • Power supply:
    Rechargeable Li-ion battery and AC outlet power option
  • Scales:
    250 scales
  • Temperature compensation range:
    10...40 °C (50...104 °F)