Order Entry
Belgium
Orders LinkContactUsLinkComponent
Lovibond® SD40 Multi Meter, Handheld

Lovibond® SD40 Multi Meter, Handheld

SD40 Multi
Catalog # LOVI720040
Supplier:   Lovibond
Lovibond® SD40 Multi Meter, Handheld
Catalog # LOVI720040
Supplier:  Lovibond
Lovibond® SD40 Multi Meter, Handheld
Lovibond® SD40 Multi Meter, Handheld

SD40 Multi

Price
206,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

The SD40 Multi meter is a compact, easy to use handheld meter for the measurement of pH/Con/TDS/Salt/T.

  • Transparent lid for easy maintenance and storage
  • Ideal lid design for a stable stand
  • Display colours and symbols for easy use
  • New multi-parameter models for individual measurement requirements
  • Robust plastic case with hanger

With its waterproof housing, the Pocket Tester is ideal for regular checks in industrial plants or swimming pools.

Plus, the transparent lid not only provides a secure stand for the Pocket Tester in daily use, but also makes it easier to maintain the electrodes. A three-colour LED lighting system - blue, green and red - and new display icons make the tester easy to use and intuitive to operate. The use of energy saving LEDs and 4×AAA batteries greatly extends the life of the new Pocket Tester range to up to 2000 hours.

With the included lanyard, each SD Series tester can be worn comfortably around the neck or attached to a pocket, waistband or belt with the clip on the head of the tester, allowing it to be taken anywhere without tiring the hands.

Delivery information: Meter is supplied with four batteries, lanyard, robust carrying case, pH buffer tablets 4/7/10 and instruction manual.

Specifications
  • Description:
    SD40 Multi
  • IP rating:
    IP 67
  • Conductivity resolution:
    Auto Scale – Con: 0,1 µS/cm, 1 µS/cm, 0,01 mS/cm
  • Power supply:
    4×1,5 V AAA batteries (>2000 hours continuous use)
  • Salinity resolution:
    0,01 ppt
  • TDS accuracy:
    ±1% FS
  • Conductivity accuracy:
    ±1% FS
  • Temperature range:
    0...50 °C
  • Salinity range:
    0,01 - 10 g/L ppt
  • Operating temperature range:
    0…60 °C
  • pH range:
    0,00 - 14,00
  • Dimensions:
    180×40×40 mm
  • Calibration:
    1, 2 or 3 point (with auto-recognition (NIST / IUPAC) and Con: 84 µS/cm/1413 µS/cm/12,88 mS/cm)
  • Temperature compensation:
    Automatic
  • Temperature resolution:
    0,1 °C
  • TDS resolution:
    Auto Scale – TDS: 0,1 ppm, 1 ppm, 0,01 ppt
  • Temperature accuracy:
    ±0,5 °C
  • Salinity accuracy:
    ±1% FS
  • Weight:
    151 g
  • Cat. No.:
    LOVI720040
  • Operating humidity range:
    0 - 80% rel. humidity (non-condensing)
  • pH accuracy:
    pH: ±0,01 (±1 digit), pH (mV): ±0,2% FS
  • pH resolution:
    0,01
  • Conductivity range:
    0,01 - 20,00 mS/cm
  • TDS range:
    0,01 - 20,00 ppt
  • Display:
    3 coloured backlight LCD (blue: measuring, green: calibration, red: alarm)
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Measuring beaker, 100 ml
    Measuring beaker, 100 ml
    634-8766
    Log in for availability
    1 * 1 ST - 7,90 €

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer set
    pH buffer set
    LOVI721250
    Log in for availability
    1 * 1 SET - 19,30 €

Product Information

  • Catalog #
    Availability
    Price
  • Replacement electrode for SD40 Multi
    Replacement electrode for SD40 Multi
    LOVI720045
    Log in for availability
    1 * 1 ST - 106,00 €

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer, 7,00 (25 °C), 90 ml
    pH buffer, 7,00 (25 °C), 90 ml
    LOVI721248
    Log in for availability
    1 * 90 mL - 7,20 €

Product Information

  • Catalog #
    Availability
    Price
  • pH buffer, 4,00 (25 °C), 90 ml
    pH buffer, 4,00 (25 °C), 90 ml
    LOVI721247
    Log in for availability
    1 * 90 mL - 7,20 €

Product Information

  • Catalog #
    Availability
    Price
  • Storage solution for pH/ORP electrodes
    Storage solution for pH/ORP electrodes
    LOVI726404
    Log in for availability
    1 * 100 mL - 13,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Buffer pH 10,01 (25 °C) blue NIST
    Buffer pH 10,01 (25 °C) blue NIST
    LOVI721249
    Log in for availability
    1 * 90 mL - 7,20 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.