Orders LinkContactUsLinkComponent
Pipe Marker, Compressed Air
Pipe Marker, Compressed Air
Pipe marker, compressed air, Polyester, 150×12 mm
Catalog # BRDYN004705
Supplier:   Brady
Pipe Marker, Compressed Air
Catalog # BRDYN004705
Supplier:  Brady
Pipe Marker, Compressed Air
Pipe Marker, Compressed Air
Pipe marker, compressed air, Polyester, 150×12 mm
Price
12,30 €
Log in for availability
1 * 1 ST
Product Details & Documents
Individual pipe markers on map with perforated arrows, without pictogram.
  • Material: Self-adhesive laminated polyester
  • Colour: White on blue
  • Weatherproof; UV and chemical resistant
Available in multiple sizes and on or without a liner.
Specifications
  • Description:
    Pipe marker, compressed air
  • Dimensions:
    150×12 mm
  • Material:
    Polyester