Order Entry
Belgium
ContactUsLinkComponent
RPMI 1640 Medium without L-Glutamine, L-Serine, Methionine, HEPES (Powder)
RPMI 1640 Medium without L-Glutamine, L-Serine, Methionine, HEPES (Powder)
N° de catalogue USBIR8999-16-10L
Fournisseur:  US Biological
Numéro CAS:  
RPMI 1640 Medium without L-Glutamine, L-Serine, Methionine, HEPES (Powder)
N° de catalogue USBIR8999-16-10L
Fournisseur:  US Biological
Numéro CAS:  

Spécifications

  • Media Format:
    Powder
  • Pk:
    1
  • Description:
    RPMI 1640 medium without L-glutamine, L-serine, methionine, HEPES (powder)
  • Size:
    10 L
  • HEPES:
    Yes
  • Phenol red:
    Yes
  • L-Glutamine:
    No
  • Glucose level:
    High
  • Media Base:
    RPMI 1640 - Roswell Park Memorial Institute Medium
  • Storage temperature:
    Store powdered media at RT. Stable for 12 months after receipt. Opened bottles should be capped tightly and kept in a dark, low humidity environment. Prepared media should be kept at 4 °C and used within a short period of time.
  • Shipping temperature:
    Powder, RT

Spécifications

À propos de cet article

RPMI-1640 was developed by Moore, et. al. at Roswell Park Memorial Institute, hence the acronym RPMI. The formulation is based on the RPMI-1630 series of media utilizing a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins. RPMI-1640 medium has been used for the culture of human normal and neoplastic leukocytes. RPMI-1640, when properly supplemented, has demonstrated wide applicability for supporting growth of many types of cultured cells, including fresh human lymphocytes in the 72 hour phytohemaglutinin (PHA) stimulation assay.

  • Appearance: Pale pink, homogenous, free flowing powder
  • Solubility: Magenta to red, clear, complete
  • Endotoxin: ≤1 EU/ml

Directions per Liter: Dissolve 10,09 g in 900 ml of ddH₂O, stirring until completely solubilised. Do NOT heat. Add 2 g sodium bicarbonate with stirring. Adjust pH of the medium to 0,1 to 0,3 pH units below the desired pH. Add additional water to bring the solution to final volume. Filter-sterilise using a 0,22 µm membrane. Aliquot into sterile containers.