Orders LinkContactUsLinkComponent
Sample Containers, Sterile
 :   BURKLE
Sample Containers, Sterile
 :  BURKLE
Sample Containers, Sterile
Sample Containers, Sterile
Sample box with tightly closing sealing cover.
  • Tightly closing sample container
  • With attached, vertically fixable snap-on lid
  • PP, transparent
The lid can be opened with one hand and its hinge angle is 90°. Liquid/condensate in the lid runs into an absorbing groove integrated in the lid, nothing is lost, and nothing can drip. The box is completely tight, has no crevices, and is therefore easy to fill and can be emptied without residues. It is autoclavable and suitable for deep freezing.
Product Family Options
  • Capacity
    Description
    Ext.Ø
    Height
    Sterile
  • 50 ml
    Graduated
    31 mm
    78 mm
    +
    1 * 650 ST
    1 * 650 ST
    New Product
    New Product
  • 90 ml
    Graduated to 80 ml
    43 mm
    79 mm
    +
    1 * 350 ST
    1 * 350 ST - 306,00 €
    306,00 €
    1 * 350 ST
       306,00 €
  • 300 ml
    Graduated
    75 mm
    88 mm
    +
    1 * 240 ST
    1 * 240 ST - 568,00 €
    568,00 €
    1 * 240 ST
       568,00 €