Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant (from E.coli)
Human Recombinant (from E.coli)
Catalog # NOVUNBP2-35298-1MG
Supplier:  Novus Biologicals
CAS Number:  
Human Recombinant (from E.coli)
Catalog # NOVUNBP2-35298-1MG
Supplier:  Novus Biologicals
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Storage conditions:
    Store at −20...−70 °C as supplied. After reconstitution, store at 2...8 °C for 1 month and at −20...−70 °C for long term storage. Avoid repeated freeze-thaw cycles.
  • Carrier-free:
    Y
  • Purity:
    >97% pure by SDS-PAGE and HPLC.

Specifications

About this item