Human Recombinant Rhesus Macaque (from E.coli)
Catalog # NOVUNBP2-61347-1MG
Supplier: Novus Biologicals
CAS Number:
Specifications
- Pk:1 mg
- Storage conditions:Store at −20...−70 °C as supplied. After reconstitution, store at 2...8 °C for 1 month and at −20...−70 °C for long term storage. Avoid repeated freeze-thaw cycles.
- Purity:>97% pure by SDS-PAGE and HPLC.