Order Entry
Belgium
ContactUsLinkComponent
Anti-Mouse Monoclonal Antibody [clone: rMT1/410] (FITC)
Anti-Mouse Monoclonal Antibody [clone: rMT1/410] (FITC)
Catalog # NOVUNBP3-08824F
Supplier:  Novus Biologicals
Anti-Mouse Monoclonal Antibody [clone: rMT1/410] (FITC)
Catalog # NOVUNBP3-08824F
Supplier:  Novus Biologicals

Specifications

  • Antibody type:
    Primary
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: rMT1/410
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human