ContactUsLinkComponent
Glass Inserts for 96 Well Plates, Microsolv tech corp
Glass Inserts for 96 Well Plates, Microsolv tech corp
Catalog # MICS95096-710
Supplier:  MICROSOLV
Glass Inserts for 96 Well Plates, Microsolv tech corp
Catalog # MICS95096-710
Supplier:  MICROSOLV
Specifications
  • Capacity:
    1,00 ml
  • Description:
    Glass inserts for 96 well plates
  • Material:
    Borosilicate glass
  • Pk:
    250
Specifications
About this item
Glass inserts for 96 well plates, 1 ml, replacements. These borosilicate glass inserts are 1 ml and are replacements for 95096-1CP or 95096-3CP.
Ordering information: Plates and mats are sold separately.