Order Entry
Belgium
ContactUsLinkComponent
QUANTOFIX® Test Strips for Semi-quantitative Determinations of pH and Nitrite in Water Analyses
QUANTOFIX® Test Strips for Semi-quantitative Determinations of pH and Nitrite in Water Analyses
Catalog # MANA91364
Supplier:  MACHEREY-NAGEL
CAS Number:  
QUANTOFIX® Test Strips for Semi-quantitative Determinations of pH and Nitrite in Water Analyses
Catalog # MANA91364
Supplier:  MACHEREY-NAGEL
CAS Number:  

Specifications

  • Chemical method:
    Nitrite - Griess reaction, pH - mixed indicator
  • Pk:
    100
  • Dimensions:
    40×40×120 mm
  • Storage temperature:
    4...30 °C
  • Shelf life (months):
    30
  • Weight:
    44,2 g
  • Parameter(s):
    Nitrite, pH

Specifications

About this item

Semi-quantitative test strips for the determination of nitrite and pH. Ideal for rapid and easy analysis of nitrite and pH directly on-site. No maintenance or calibration is required.

  • Nitrite - 0, 1, 5, 10, 20, 40, 80, 100 mg/l NO₂⁻
  • pH - 8,0, 8,2, 8,4, 8,6, 8,8, 9, 9,2, 9,4, 9,6, 9,8, 10
  • Color change nitrite - white  to  red-violet
  • Color change pH - yellow-orange to violet-red