Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

CryoStor® CS2 Cell Freezing Medium

CryoStor® CS2, 100 ml, Human, mouse, non human primate, other, rat; Animal component-free, serum-free, 100 mL
Catalog # STMC07932
Marketsource
CryoStor® CS2 Cell Freezing Medium
Catalog # STMC07932
Marketsource
CAS Number:  
CryoStor® CS2 Cell Freezing Medium
CryoStor® CS2 Cell Freezing Medium

CryoStor® CS2, 100 ml, Human, mouse, non human primate, other, rat; Animal component-free, serum-free, 100 mL

CAS Number:  
Price
438,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

Animal component-free, defined cryopreservation medium with 2% DMSO.

  • Ready-to-use
  • Serum-free and protein-free
  • Animal component-free
  • cGMP manufactured with USP grade/highest-quality components
  • FDA master file
  • Sterility, endotoxin, and cell-based quality control testing

Maximise post-thaw cell recovery and viability following cryopreservation at low temperatures (−80 to −196 °C) with ready-to-use CryoStor® CS2. Serum-free, animal component-free, and cGMP-manufactured, this defined medium provides a safe, protective environment for cells and tissues during freezing and thawing processes and storage. CryoStor® CS2 is formulated with USP-grade components and contains 2% DMSO.

 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    CryoStor® CS2, 100 ml
  • Format:
    Human, mouse, non human primate, other, rat; Animal component-free, serum-free

Specifications