CryoStor® CSB Cryopreservation Medium
About this item
Animal component-free and protein-free cryopreservation formulation.
- Serum-free and chemically defined
- USP grade/multi-compendial/high-quality components
- cGMP-manufactured
- Sterility, endotoxin, and cell-based release testing
- FDA master file
Reduce the level of cryopreservation-induced delayed-onset cell death and maximise post-thaw cell recovery, viability, and function following cryopreservation, with CryoStor® CSB, an intracellular-like base formulation from the CryoStor® preservation media series. Serum-free, animal component-free, and cGMP-manufactured, this chemically defined medium is sterile and tested for endotoxins and cell-based release. CryoStor® CSB is formulated with USP-grade components and does not contain DMSO.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
3 Options Available Below
Product Details & Documents
Animal component-free and protein-free cryopreservation formulation.
- Serum-free and chemically defined
- USP grade/multi-compendial/high-quality components
- cGMP-manufactured
- Sterility, endotoxin, and cell-based release testing
- FDA master file
Reduce the level of cryopreservation-induced delayed-onset cell death and maximise post-thaw cell recovery, viability, and function following cryopreservation, with CryoStor® CSB, an intracellular-like base formulation from the CryoStor® preservation media series. Serum-free, animal component-free, and cGMP-manufactured, this chemically defined medium is sterile and tested for endotoxins and cell-based release. CryoStor® CSB is formulated with USP-grade components and does not contain DMSO.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
Product Information
- FormatPkAvailabilityPrice
- STMC100-0237CryoStor® CSB, 100 mlCryoStor® CSB, 100 mlHuman; Animal component-free, chemically defined, serum-free100 mLLog in for availability1 * 100 mL686,00 €Specifications:MarketsourceMarketsource
- STMC100-0238CryoStor® CSB, 500 mlCryoStor® CSB, 500 mlHuman; Animal component-free, chemically defined, serum-free1 KITLog in for availability1 * 1 KIT3 476,00 €Specifications:MarketsourceMarketsource
- STMC100-0239CryoStor® CSB, 1000 mlCryoStor® CSB, 1000 mlHuman; Animal component-free, chemically defined, serum-free1 KITLog in for availability1 * 1 KIT7 134,00 €Specifications:MarketsourceMarketsource