Anti-S6 Ribosomal Protein (Ser235/Ser236) Phosphate Rabbit Polyclonal Antibody
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: S6 Ribosomal Protein (Ser235/Ser236) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Monkey, S. cerevisiae
Specifications
- Catalog No:
- CSTA2211L
- CSTA2211S
- Antigen symbol:
- S6 Ribosomal Protein (Ser235/Ser236) phosphate
- S6 Ribosomal Protein (Ser235/Ser236) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- S. cerevisiae, Monkey, Human, Rat, Mouse
- S. cerevisiae, Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CRRLssLRASTSKSES
- CRRLssLRASTSKSES
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together