Order Entry
Belgium
ContactUsLinkComponent
 
Anti-Yes (Tyr426) phosphate Rabbit Monoclonal Antibody [clone: D49G4]
 
Anti-Yes (Tyr426) phosphate Rabbit Monoclonal Antibody [clone: D49G4]

 

  • Catalog No:
  • CSTA6943T
  • CSTA6943S
  • Antigen symbol:
  • Yes (Tyr426) phosphate
  • Yes (Tyr426) phosphate
  • Antigen synonyms:
  • Src (Tyr419) phosphate|Yes (Tyr426) phosphate|Lyn (Tyr397) phosphate|Fyn (Tyr420) phosphate|Lck (Tyr394) phosphate|Hck (Tyr411) phosphate
  • Src (Tyr419) phosphate|Yes (Tyr426) phosphate|Lyn (Tyr397) phosphate|Fyn (Tyr420) phosphate|Lck (Tyr394) phosphate|Hck (Tyr411) phosphate
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • D49G4
  • D49G4
  • Reactivity:
  • Monkey, Human, Rat, Mouse
  • Monkey, Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Sequence:
  • RLIEDNEyTARQGAKC
  • RLIEDNEyTARQGAKC
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • Ambient
  • Ambient

 

Frequently Bought Together