Anti-NKCC1 Rabbit Polyclonal Antibody
Catalog # CSTA14581S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:NKCC1
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Rat,Mouse
- Western blot:Yes
- Sequence:CLGRPLGPTPSQSRFQVDPVSENAGR
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NKCC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat