ContactUsLinkComponent
Mouse Reactive Inflammasome Antibody Sampler Kit
Mouse Reactive Inflammasome Antibody Sampler Kit
Catalog # CSTA20836T
Supplier:  Cell Signaling
Mouse Reactive Inflammasome Antibody Sampler Kit
Catalog # CSTA20836T
Supplier:  Cell Signaling
Specifications
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    Ambient
  • Pk:
    1 KIT
Specifications
About this item
The Mouse Reactive Inflammasome Antibody Sampler Kit provides an economical means of detecting multiple inflammasome components. The kit includes enough antibodies to perform at least two western blot experiments with each primary antibody.
Each antibody in the Mouse Reactive Inflammasome Antibody Sampler Kit detects endogenous levels of its target protein. Caspase-1 (E2Z1C) Rabbit mAb detects endogenous levels of full-length mouse caspase-1; this antibody detects pro-caspase-1 and the p10 subunit of activated caspase-1. Cleaved Caspase-1 (Asp296) (E2G2I) Rabbit mAb detects endogenous levels of mouse caspase-1 only when cleaved at Asp296. A non-specific band is detected at 70 kDa in some cells. Cleaved-IL-1β (Asp117) (E7V2A) Rabbit mAb (Mouse Specific) recognises endogenous levels of mouse IL-1β protein only when cleaved at Asp117.
Delivery: Kit includes NLRP3 (D4D8T) Rabbit mAb (CSTA15101) 20 µl, AIM2 Antibody (CSTA63660) 20 µl, ASC/TMS1 (D2W8U) Rabbit mAb (CSTA67824) 20 µl, Cleaved-IL-1β (Asp117) (E7V2A) Rabbit mAb (CSTA63124) 20 µl, IL-1β (D6D6T) Rabbit mAb (CSTA31202) 20 µl, Cleaved Caspase-1 (Asp296) (E2G2I) Rabbit mAb (CSTA89332) 20 µl, Caspase-1 (E2Z1C) Rabbit mAb (CSTA24232) 20 µl, Anti-rabbit IgG 20 µl, HRP-linked Antibody (CSTA7074) 100 µl.