Anti-Casp1 (p20) cleaved Rabbit Monoclonal Antibody [clone: D57A2]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: Casp1 (p20) cleaved
Clonality: Monoclonal
Clone: D57A2
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- CSTA4199S
- CSTA4199T
- Antigen symbol:
- Casp1 (p20) cleaved
- Casp1 (p20) cleaved
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D57A2
- D57A2
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CGGQAGRGDSPGVVWFKD
- CGGQAGRGDSPGVVWFKD
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together