Order Entry
Belgium
ContactUsLinkComponent
Anti-cre recombinase Rabbit Monoclonal Antibody [clone: D7L7L]
Anti-cre recombinase Rabbit Monoclonal Antibody [clone: D7L7L]
Supplier:  Cell Signaling
Certificates
Anti-cre recombinase Rabbit Monoclonal Antibody [clone: D7L7L]
Supplier:  Cell Signaling

Specifications

  • Catalog No:
  • CSTA15036S
  • CSTA15036T
  • Antigen symbol:
  • cre recombinase
  • cre recombinase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Clone:
  • D7L7L
  • D7L7L
  • Reactivity:
  • Multiple
  • Multiple
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Sequence:
  • CSNLLTVHQNLPALPVDATSDEVRKNL
  • CSNLLTVHQNLPALPVDATSDEVRKNL
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • Ambient
  • Ambient

Specifications

Frequently Bought Together