Anti-NMDAR1 Rabbit Monoclonal Antibody [clone: D65B7]
Catalog # CSTA5704S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:NMDAR1
- Clonality:Monoclonal
- Clone:D65B7
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CLDRPEERITGINDPRLRNPSDK
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: NMDAR1
Clonality: Monoclonal
Clone: D65B7
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat