Anti-RIPK1 Rabbit Monoclonal Antibody [clone: D94C12]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: RIPK1
Clonality: Monoclonal
Clone: D94C12
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Hamster, Monkey
Specifications
- Catalog No:
- CSTA3493T
- CSTA3493S
- Antigen symbol:
- RIPK1
- RIPK1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D94C12
- D94C12
- Reactivity:
- Hamster, Monkey, Human, Rat, Mouse
- Hamster, Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CNGGTLYYMAPEHLNDVNAKPTEKSD
- CNGGTLYYMAPEHLNDVNAKPTEKSD
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together