Anti-NPY Rabbit Monoclonal Antibody [clone: D7Y5A]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: NPY
Clonality: Monoclonal
Clone: D7Y5A
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- CSTA11976S
- CSTA11976T
- Antigen symbol:
- NPY
- NPY
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D7Y5A
- D7Y5A
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Sequence:
- CYPSKPDNPGEDAPAED
- CYPSKPDNPGEDAPAED
- Storage temperature:
- –20 °C
- –20 °C
- Shipping temperature:
- Ambient
- Ambient
Specifications
Frequently Bought Together