Anti-AMPK-alpha1 (Thr183) phosphate Rabbit Monoclonal Antibody [clone: '40H9]
Supplier: Cell Signaling
Certificates
About this item
Type: Primary Antibodies
Antigen: AMPK-alpha1 (Thr183) phosphate
Clonality: Monoclonal
Clone: '40H9
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat,Hamster,Monkey,D. melanogasterm,S. cerevisiae
Specifications
- Catalog No:
- CSTA2535S
- CSTA2535T
- Antigen symbol:
- AMPK-alpha1 (Thr183) phosphate
- AMPK-alpha1 (Thr183) phosphate
- Antigen synonyms:
- AMPK-alpha2 (Thr172) phosphate
- AMPK-alpha2 (Thr172) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 40H9
- 40H9
- Reactivity:
- S. cerevisiae, Hamster, Monkey, Human, D. melanogasterm, Rat, Mouse
- S. cerevisiae, Hamster, Monkey, Human, D. melanogasterm, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- SDGEFLRtSCGSPNY
- SDGEFLRtSCGSPNY
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together