Orders LinkContactUsLinkComponent
Bump Cap Micro Peak, AirTech
Bump Cap Micro Peak, AirTech
Supplier:   Portwest
Bump Cap Micro Peak, AirTech
Supplier:  Portwest
Certificates
Bump Cap Micro Peak, AirTech
2 Options Available Below
Product Details & Documents
Innovative design bump cap with maximised ventilation thanks to the side mesh and ventilation slots in the plastic shell.
  • Micro peak (25 mm) for better visibility and for use in confined spaces
  • Cooling mesh fabric for increased breathability
  • Inner ABS shell and soft pad for shock absorption
  • Reflective piping for increased visibility
  • Retail bag aids presentation for retail sales
Modern textile design with reflective piping for enhanced visibility. Micro peak makes the bump cap suitable for indoor use or applications in confined spaces. Equipped with a buckle size adjustment for easy fitting.
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • PRTWPS69BKR
    AirTech bump cap micro peak
    AirTech bump cap micro peak
    Black
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,90 €
    19,90 €
    1 * 1 ST
       19,90 €
  • PRTWPS69NAR
    AirTech bump cap micro peak
    AirTech bump cap micro peak
    Navy
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,90 €
    19,90 €
    1 * 1 ST
       19,90 €