Orders LinkContactUsLinkComponent
Height Endurance Visor
Height Endurance Visor
Height endurance visor, Clear
Catalog # PRTWPA03CLR
Supplier:   Portwest
Height Endurance Visor
Catalog # PRTWPA03CLR
Supplier:  Portwest
Height Endurance Visor
Height Endurance Visor
Height endurance visor, Clear
Price
40,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Panoramic wrap-around visor for Height Endurance helmets range. It allows the user to wear over prescription glasses.
  • One-piece curved lens with moulded nose bridge
  • Light and robust
  • Anti-scratch coating for added durability
  • Anti-fog coating for additional comfort to the wearer
  • Retail box aids presentation for retail sales
Compatible with Height Endurance Helmets and suitable for wearing over most prescription glasses, this visor features an anti-scratch and anti-fog treatment for added durability.
Certifications: EN 170 (2C-1.2 ).
Specifications
  • Colour:
    Clear
  • Description:
    Height endurance visor