Anti-SLC7A11 Rabbit Monoclonal Antibody [clone: D2M7A]
Catalog # CSTA12691S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:SLC7A11
- Clonality:Monoclonal
- Clone:D2M7A
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CIKGQTQNFKDAFSGRDSSITR
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl