Order Entry
Belgium
ContactUsLinkComponent
Human Recombinant alpha-Amylase 2B (from Cells)
Human Recombinant alpha-Amylase 2B (from Cells)
Catalog # PRSI92-341
Supplier:  ProSci Inc.
CAS Number:  
Human Recombinant alpha-Amylase 2B (from Cells)
Catalog # PRSI92-341
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    0,05 mg
  • Enzyme type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt, please minimise freeze - thaw cycles
  • Endotoxin content:
    <0,1 ng/µg (1 IEU/µg) as determined by LAL test
  • Enzyme name:
    alpha-Amylase 2B
  • Enzyme synonyms:
    1,4-alpha-D-glucan glucanohydrolase 2B, Carcinoid alpha-amylase, HXA, AMY2, AMY3, AMY2B
  • Purity:
    >95% as determined by reducing SDS-PAGE
  • Molecular weight:
    56,9 kD
  • Sequence:
    Gln16-Leu511
  • Formulation:
    Supplied as a 0,2 µm filtered solution of 20 mM PB, 150 mM NaCl,pH 7,4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
  • Tested applications:
    Biological assays

Specifications

About this item

Alpha-amylase 2B is an enzyme that in humans is encoded by the AMY2B gene.

This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag

Alpha-amylase 2B is a secreted protein and belongs to the glycosyl hydrolase 13 family. It hydrolyzes 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. This gene encodes an amylase isoenzyme produced by the pancreas. Diseases associated with AMY2B include pancreatitis, and thyroiditis, and among its related super-pathways are Metabolic pathways and MPS IIIC - Sanfilippo syndrome C.