Human Recombinant Sulfotransferase 2B1 (from E. coli)
Catalog # PRSI92-053
Supplier: ProSci Inc.
New Product
CAS Number:
Specifications
- Pk:0,05 mg
- Enzyme type:Recombinant
- Source:E. coli
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Store at −20 °C, stable for 6 months after receipt, please minimise freeze - thaw cycles
- Endotoxin content:<0,1 ng/µg (1 IEU/µg) as determined by LAL test
- Enzyme name:Sulfotransferase 2B1
- Enzyme synonyms:Sulfotransferase Family Cytosolic 2B Member 1, ST2B1, Sulfotransferase 2B1, Alcohol Sulfotransferase, Hydroxysteroid Sulfotransferase 2, SULT2B1, HSST2
- Purity:>95% as determined by reducing SDS-PAGE
- Molecular weight:36,6 kD
- Sequence:Met1-Glu311
- Formulation:Supplied as a 0,2 µm filtered solution of 20 mM Tris, 0,1 M NaCl, 1 mM EDTA, pH 8,0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 µg/ml. Dissolve the lyophilised protein in ddH₂O. Please aliquot the reconstituted solution to minimise freeze-thaw cycles.
- Tested applications:Used for biological assays
Specifications
About this item
Sulfotransferase Family Cytosolic 2B Member 1(SULT2B1) is a member of the Sulfotransferase 1 family.
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
SULT2B1 is highly expressed in the placenta, prostate, and trachea with lower expression levels in the small intestine and lung. SULT2B1 catalyses the sulfate conjugation of numerous hormones, neurotransmitters, drugs, and xenobiotic compounds. Sulfonation enhances the water solubility of molecules, and therefore their renal excretion, however SULT2B1 can also result in bioactivation to form active metabolites.