Anti-DARPP-32 Rabbit Monoclonal Antibody [clone: 19A3]
CSTA2306S
- Antibody type:Primary
- Antigen symbol:DARPP-32
- Clonality:Monoclonal
- Clone:19A3
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Rat,Mouse
- Western blot:Yes
- Sequence:CGQGLEGPWERPPPLDESERDG
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: DARPP-32
Clonality: Monoclonal
Clone: 19A3
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse;Rat