Anti-ULK1 (Ser758) phosphate Rabbit Monoclonal Antibody [clone: D7O6U]
Type: Primary
Antigen: ULK1 (Ser758) phosphate
Clonality: Monoclonal
Clone: D7O6U
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey
- Catalog No:
- CSTA14202T
- CSTA14202S
- Antigen symbol:
- ULK1 (Ser758) phosphate
- ULK1 (Ser758) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- D7O6U
- D7O6U
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Sequence:
- CPVVFTVGsPPSGST
- CPVVFTVGsPPSGST
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Ambient
- Ambient
Frequently Bought Together