Spécifications
- Description:SD30 EC pH Pocket tester
- Model:SD30 EC
- Pk:1
Spécifications
À propos de cet article
SD30 EC With its waterproof housing, the pocket tester is ideal for regular checks in industrial plants or swimming pools.
- Accuracy Con/TDS/Salt: ± 1 % FS
- Temperature: ± 0.5 °C
- Display 3 coloured backlit LCD(blue: measurement, green: calibration, red: alarm)
- Auto-OFF able
- Calibration 1 or 2 points calibration for auto mode Standard:1413 μS/cm or Standard: 12.88 mS/cm up to 2 points calibration for manual mode ± 50 % adjustable value
- Portable
- Temperature Compensation: Automatic
- Resolution Auto Scale - Con 0.1 µS/cm, 1 µS/cm, 0.01 mS/cm; Auto Scale – TDS: 0.1 ppm, 1 ppm, 0.01 ppt; Salt: 0.01 pp
Plus, the transparent lid not only provides a secure stand for the pocket tester in daily use, but also makes it easier to maintain the electrodes. A three-colour LED lighting system - blue, green and red - and new display icons make the tester easy to use and intuitive to operate.
The use of energy saving LEDs and 4 x AAA batteries greatly extends the life of the new Pocket Tester range to up to 2000 hours.
With the included lanyard, each SD Series tester can be worn comfortably around the neck or attached to a pocket, waistband or belt with the clip on the head of the tester, allowing it to be taken anywhere
without tiring the hands.
It can be using in chemical industry, food and beverage industry, industries others, marine industry, municipalities, NGO, oil and gas, pharmaceutical Industry, pool public, pool services.
Resolution temperature: 0,1 °C.
Calibration points: 1 to 3.
Buffer auto recognition: Con: 84 µS/cm/1413 µS/cm/12 mS/cm.
Calibration indicator: Yes.
Stability indicator: Yes.
Auto-hold function: Yes.
Alarm limits: Yes.
Environmental conditions: 0 to 60 °C/0 to 80% rel.humidity (non-condensing).
Dimensions: 40×40×180 mm.
Weight 151 g.
Weight with Packaging: 268 g.